missing translation for 'onlineSavingsMsg'
Learn More

Standard Buffer Solution, SI Analytics™

Product Code. 11954779
Click to view available options
Quantity:
60 Ampules
This item is not returnable. View return policy
This item is not returnable. View return policy

SI Analytics™ Standard Buffer Solution pH 6.87 resists changes in pH when small quantities of an acid are added to it.

  • Includes manufacturer certificate
  • Mixture, consisting of the following components: Water, disodium hydrogen phospate, potassium dihydrogen phosphate
  • The product does not contain any relevant quantities of materials with critical values that have to be monitored at the workplace
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 100°C
Color Colorless
Density 1g/cm3
pH 6.87
Physical Form Liquid
Quantity 60 Ampules
Formulation Water, disodium hydrogen phospate, potassium dihydrogen phosphate
Odor Odorless
Packaging FIOLAX™ Ampules
Sterility Sterile
Vapor Pressure 23hPa
Solubility Information Fully miscible in water
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'