Learn More
Koettermann™ Standard Cabinet for Acids and Bases, PP, 520x600x1920mm
Koettermann™ Standard Cabinet for Acids and Bases provides suitable storage to accommodate a variety of laboratory needs, but is especially suited for storing acids and bases.
Brand:  Koettermann™ 377-00002
 View more versions of this product
Product Code. 15518545
  • 4470.00 € / Each
Explore more special offers
Description
Description
  • Tall, Acid/Base Cabinet
  • Two doors
  • Four drawers
Specifications
Specifications
520 x 600 x 1920 mm
Acids and Bases
2
Right-hinged
4 Drawers, No Fan
Acids and Bases Cabinet