Learn More
Corning™ Standard Conical Bioprinting Nozzles
These Standard Conical Bioprinting Nozzles are designed fo use with Matribot™ Bioprinter. They are made from polypropylene, have a female Luer connector, and are available in 22G, 25G, and 27G.
Brand:  Corning™ 6166
 View more versions of this product
Product Code. 17780789
  • 58.35 € / Pack of 50
Estimated Shipment: 16-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Made of polypropylene
  • Feature a female Luer connector
  • Available in 22 G, 25 G, and 27 G.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Red
Polypropylene
Feature Female Luer Connector
1.26 in.
50 Nozzles
Bioprinting Nozzle
250 μm
25 G
32 mm