Learn More
Koettermann™ Standard Hanging Cabinet, 1500x480x366
Koettermann™ Standard Hanging Cabinet, 1500x480x366 provides suitable storage to accommodate a variety of laboratory needs.
Brand:  Koettermann™ 307-00207
Product Code. 15733122
  • 892.00 € / Each
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Wall-mounted cabinet
  • Three doors
  • One shelf
  • Lockable
  • Side panels
Specifications
Specifications
Wall-mounted Cabinet
Yes
1
350 x 1500 x 480 mm
3