missing translation for 'onlineSavingsMsg'
Learn More

Koettermann™ Standard Individual Table, C-Frame, 1200x750x750mm

Koettermann™ Standard Individual Table, C-Frame, 1200x750x750mm is a free-standing table with a durable worktop and C-type legs.

Brand:  Koettermann™ 505-00062

Product Code. 15548555

  • 1450.00 € / Each
Estimated Shipment: 16-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Table with C-type legs
  • Trespa Toplab Plus worktop
Specifications

Specifications

Table
750 mm
C-type legs
750 mm
Trespa Toplab Plus Worktop
1200 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Koettermann™ Standard Individual Table, C-Frame, 1200x750x750mm > Includes: Trespa Toplab Plus Worktop

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.