missing translation for 'onlineSavingsMsg'
Learn More

Koettermann™ Standard Individual Table, C-Frame, 900x750x750mm

Koettermann™ Standard Individual Table, C-Frame, 900x750x750mm is a free-standing table with a durable worktop and C-type legs.

Brand:  Koettermann™ 505-00055

Product Code. 15578545

  • 995.00 € / Each
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Table with C-type legs
  • TopResist worktop
Specifications

Specifications

Table
750 mm
C-type legs
750 mm
TopResist Worktop
900 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers