missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tenak™ Standard Rack for Upright Freezer, 130 mm Box Height
Standard Rack fits into any upright freezer and can be used with all standard cryoboxes.
Brand: Tenak™ TE23330
This item is not returnable.
View return policy
Description
- Measure the inside height and depth of your freezer when choosing the right rack
- Racks can be used with all freezer brands
- Constructed of stainless steel
Specifications
2 x 4 | |
Stainless Steel | |
Upright Freezers | |
130 mm (Box) |
Boxes | |
8 | |
560 x 140 x 268 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction