missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Standard RC Dialysis Tubing

Product Code. p-4394892
Click to view available options
Diameter (Metric):
6.4 mm
16 mm
20.4 mm
29 mm
48 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11465859

Brand: Spectrum™ 132709

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Multipurpose dialysis tubing is ideal for general dialysis for a variety of applications and volumes

  • Membrane grade
  • Membrane type: RC
  • Molecular weight cut-off: 12 to 14K

Specifications

Product Type Dialysis Membrane
MWCO 12 to 14 kDa
Material Regenerated Cellulose
Diameter (Metric) 48 mm
Diameter (English) 1.88 in.
Length (English) 50 ft.
Length (Metric) 15 m
Capacity (Metric) 18 mL
Quantity 50 Pack
For Use With (Application) Some tubing rolls do not contain glycerin and do not require soaking prior to use
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.