missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Standard Sample Tube, 3.24mm ID

For use with MKII Magnetic Susceptibility Balances

75.58 € - 1137.66 €

Specifications

Type Standard Sample Tube
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11480400
View Documents
Thermo Scientific Chemicals
014349.KL
20 Pack
1137.66 €
Pack of 20
Please to purchase this item. Need a web account? Register with us today!
15431067
View Documents
Thermo Scientific Chemicals
014349.KJ
5 Pack
324.62 €
Pack of 5
Please to purchase this item. Need a web account? Register with us today!
11470400
View Documents
Thermo Scientific Chemicals
014349.KG
1 Each
75.58 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • I.D.: 3.24mm
Specifications

Specifications

Standard Sample Tube
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Standard Sample Tube, 3.24mm ID

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.