Learn More
Macherey-Nagel™ Standard Silica 60 Adsorbents
Brand:  Macherey-Nagel™ 815380.25
Product Code. 12760264
  • 1185.00 € / 25kg
Explore more special offers
Specifications
0.04 to 0.063 mm
Column Chromatography
Silica 60
Standard Silica 60 Adsorbent
∼60 Å
25 kg
∼500 m2/g
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Macherey-Nagel™ Standard Silica 60 Adsorbents > Particle Size: 0.04 to 0.063mm