missing translation for 'onlineSavingsMsg'
Learn More

Implen™ NanoPhotometer™ Standard Solutions

Use to control the photometric accuracy of the NanoPhotometer™ NP120, NP80, N60 and N50 (NanoVolume)

60.00 € - 300.00 €

Specifications

Type Non-toxic Potassium Hydrogen Phthalate Solution (PHP)
Storage Requirements Store at room temperature
Wavelength 280 nm
View More Specs

Products 3
Product Code Brand For Use With (Equipment) Packaging Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Packaging Price Quantity & Availability  
15713217
View Documents
Implen™
N-568-S2
NanoPhotometer N60 and NP80 2 x 0.5 mL
60.00 €
Pack of 2
Please to purchase this item. Need a web account? Register with us today!
15765805
View Documents
Implen™
N-568-S
NanoPhotometer N60 and NP80 10 x 0.5 mL
300.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
16473909
View Documents
Implen™
N-120-S
NanoPhotometer N120 5 x 1 mL (Per Solution)
300.00 €
Pack of 5
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Standard solutions for the control of photometric accuracy
  • Verifies the instrument's absorbance accuracy
  • For use with NanoPhotometer™ NP120/NP80/N60/N50
  • CAS-No: 877-24-7
Specifications

Specifications

Non-toxic Potassium Hydrogen Phthalate Solution (PHP)
Store at room temperature
280 nm
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.