missing translation for 'onlineSavingsMsg'
Learn More

Ratiolab™ Standard Waste Disposal Bags

An excellent option for the disposal of contaminated labware.

Brand:  Ratiolab™ 7001210

 View more versions of this product

Product Code. 13475879

  • 52.65 € / Pack of 100

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Manufactured from very strong polypropylene film
  • With a broad seam at the bottom
  • Autoclavable at 134°C, (bag must not be completely closed)
Specifications

Specifications

Waste Disposal Bag
32 L
Waste disposal applications
100/Pk.
Autoclavable
Single-use
Polypropylene
600 x 800 x 0.05 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.