missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stannous Chloride, Dihydrate, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Description
- Suitable for Mercury Determination
Specifications
Specifications
Physical Form | Crystals |
Quantity | 500 g |
Formula Weight | 225.63 |
Grade | Meets ACS reagent chemical requirements |
Packaging | Poly Bottle |
Specific Gravity | 1L = 2.71kg |
Chemical Name or Material | Stannous Chloride, Dihydrate |
Safety and Handling
Signal Word
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P362-Take off contaminated clothing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'