Learn More
3M™ Petrifilm™ Staph Express Count Plate
Provide Staphylococcus aureus results. S. aureus appears as distinctive red-violet colonies. Other colonies have to be checked using the Staph express disk.
121.17 € - 927.26 €
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17253774
|
3M™ Petrifilm™
7100090291 |
50 |
121.17 €
Pack of 50 |
Estimated Shipment: 05-12-2024
Log in to see stock. |
|||||
17263774
|
3M™ Petrifilm™
7100090292 |
500 |
927.26 €
Pack of 500 |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
Description
- Results in 24 Hours.
- Plates are sample-ready, eliminating all the time-consuming steps of preparing media/agar dishes.
- Unique system to facilitate the enumeration of S. aureus
- This plate has been awarded Official Methods of Analysis (OMA) #2003.07 for processed and prepared foods, #2003.8 for select dairy foods and #2003.11 for Select meat, seafood , poultry and NF VALIDATION certificates: 3M-01/9-04/03 A and 3M-01/9-04/03 B for All human foods and Pet food by AFNOR Certification.
- Food processors and contract labs worldwide trust 3M Petrifilm Plates to provide accurate, efficient, consistent results.
Specifications
Food and Beverage, Environmental Test |