Learn More
Neogen Petrifilm™ Staph Express Disk
Product Code. p-7230031
Encompass
Click to view available options
Quantity:
20
100
Unit Size:
Pack of 100
Pack of 20
Product Code. 17293754
Brand: Neogen Petrifilm™ 7100039572
This item requires a price update prior to being ordered.
Request a Price for this item here.
Used to identify Staphylococcus aureus from all suspect colonies for samples with high background flora.
  • Utilizes the convenience and efficiency of the Neogen Petrifilm™ Plate platform.
  • Proven cost effective tool for the detection of Staph. aureus.
  • Results in as few as 22 hours.
Specifications
Test Type Animal Feed, Pet Food
Quantity 20