Learn More
Novacyt - Microgen Bioproducts Staph ID
Suitable for use with clinical and veterinary samples.
Brand:  Novacyt - Microgen Bioproducts MID-69
Product Code. 17333221
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Microgen™ Staph-ID has been developed for the identification of commonly encountered Staphylococcus spp. isolated from clinical and veterinary samples. These bacteria, although commensal, are recognised as potential human pathogens e.g. S. aureus and S. epidermidis. Coagulase negative Staphylococcus spp. are common colonisers of prosthetic valves and in dwelling catheters. Gram stain (positive), catalase (positive) and latex agglutination/coagulase tests are performed as pre-tests on the isolate. Colony pigmentation is recorded and then 1-2 colonies are inoculated into the suspending medium. The suspension is inoculated into the 12 well test strip and incubated at 37°C. After 24 hours incubation the strips are read and the Nitrate and PYR reagents added. The resulting 5 digit numerical code is entered in to the software and an identification returned.
As with all Microgen™ biochemical ID systems, Staph-ID is supported by the simplest-to-use and most feature-packed Computer Identification System Software (MID60) available, delivering the most comprehensive and up-to-date databases.
Species identified:
  • Staphylococcus spp.
  • S. aureus subsp. aureus
  • S. aureus subsp. anaerobius
  • S. auricularis
  • S. caprae
  • S. capitis subsp. capitis
  • S. capitis subsp. urealyticus
  • S. carnosus
  • S. chromogenes
  • S. cohnii subsp. conhii
  • S. cohnii subsp. urealyticum
  • S. epidermidis
  • S. haemolyticus
  • S. hominis subsp. hominis
  • S. hominis subsp. novobiosepticus
  • S. hyicus
  • S. intermedius
  • S. lentus
  • S. lugdenensis
  • S. saccharolyticus
  • S. saprophyticus
  • S. schleiferi subsp. schleiferi
  • S. schleiferi subsp. coagulans
  • S. sciuri
  • S. simulans
  • S. warneri
  • S. xylosus Kocuria spp.
  • K. kristinae
  • K. rosea
  • K. carniphila Kytococcus spp.
  • Ky. sedentarius Micrococcus spp.
  • M. luteus
  • M. lylae
Specifications
Specifications
ISO13485 ISO9001
Yes
Gelatin, Malonate, Inositol, Sorbitol, Rhamnose, Sucrose, Lactose, Arabinose, Adonitol, Raffinose, Salicin, Arginine, TDA, Nitrate
Bacterial Isolate
Test Strip
20 Test strips
Monera
20 x 3 mL bottles, 20 Test strips
>99%
18 to 24 hours
2°C to 8°C
Non-waived
20 x 3 mL bottles, 20 Test strips
>99%
An Identification System for Staphylococci of Clinical Animal Health and Environmental Importance
12 per test strip
Carton with pouches and strip holding tray
Staphylococci of clinical animal health and environmental importance
Bacteria