missing translation for 'onlineSavingsMsg'
Learn More

Starch, Certified AR for Analysis, Soluble, Fisher Chemical™

Product Code. 10677612
Click to view available options
Quantity:
250 g
500 g
1 kg

C12H22O11, CAS Number-9005-25-8, starch, soluble, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose, glcalpha1-4glca, maltose, unii-15sug9ad26, starch solution, alpha-malt sugar, alpha-maltose, amylodextrin, glcalpha1-4glcalpha, 250g, CHEBI:18167, White, GUBGYTABKSRVRQ-ASMJPISFSA-N

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9005-25-8
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00082026,MFCD00132834
InChI Key GUBGYTABKSRVRQ-UHFFFAOYNA-N
Synonym alpha-maltose, maltose, starch, soluble, glcalpha1-4glca, unii-15sug9ad26, glcalpha1-4glcalpha, amylodextrin, starch solution, alpha-malt sugar, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose
PubChem CID 439341
ChEBI CHEBI:18167
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol
SMILES OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O

Specifications

Color White
pH 5 to 7
Physical Form Powder Solid
Quantity 250 g
Packaging Plastic powder jar
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.