missing translation for 'onlineSavingsMsg'
Learn More

Starch, Hydrolyzed, Ultrapure, Thermo Scientific™

Product Code. 15875358
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9005-25-8
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00082026,MFCD00132834
InChI Key GUBGYTABKSRVRQ-UHFFFAOYNA-N
Synonym alpha-maltose, maltose, starch, soluble, glcalpha1-4glca, unii-15sug9ad26, glcalpha1-4glcalpha, amylodextrin, starch solution, alpha-malt sugar, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose
PubChem CID 439341
ChEBI CHEBI:18167
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol
SMILES OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O

Specifications

Physical Form Powder
Quantity 1 kg
Packaging Plastic bottle
Chemical Name or Material Starch
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'