missing translation for 'onlineSavingsMsg'
Learn More
Starch, Potato, Powder, Thermo Scientific Chemicals Product Code.: 10105233

Starch, Potato, Powder, Thermo Scientific Chemicals

Product Code. 10105233
2.5 kg, Plastic bottle
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10105233

Brand: Thermo Scientific Acros 419690025

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9005-25-8
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00082026,MFCD00132834
InChI Key GUBGYTABKSRVRQ-UHFFFAOYNA-N
Synonym alpha-maltose, maltose, starch, soluble, glcalpha1-4glca, unii-15sug9ad26, glcalpha1-4glcalpha, amylodextrin, starch solution, alpha-malt sugar, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranoseShow MoreShow Less
PubChem CID 439341
ChEBI CHEBI:18167
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol
SMILES OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O

Specifications

Color White
pH 6 to 8 (50% aq. suspension)
Physical Form Powder
Ash 0.5% max. (900°C)
Merck Index 14, 8798
Solubility Information Solubility in water: insoluble.
Quantity 2.5 kg
Grade Extra Pure
Infrared Spectrum Authentic
Loss on Drying 20.5% max. (2 g, 130°C, 90 min)
Packaging Plastic bottle
Chemical Name or Material Starch
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466196 Certificate TypeCertificate of Analysis Date19/09/2024 Catalog Number419690025,419695000
Lot NumberA0463402 Certificate TypeCertificate of Analysis Date05/06/2024 Catalog Number419690025,419695000
Lot NumberA0456858 Certificate TypeCertificate of Analysis Date03/11/2023 Catalog Number419690025,419695000
Lot NumberA0448008 Certificate TypeCertificate of Analysis Date27/02/2023 Catalog Number419690025,419695000
Lot NumberA0400464 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number419690025,419695000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Starch, Potato, Powder, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.