missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Starch, Soluble Potato, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
216.00 € - 895.00 €
Chemical Identifiers
CAS | 9005-84-9 |
---|---|
Molecular Formula | C12H22O11 |
Molecular Weight (g/mol) | 342.297 |
InChI Key | GUBGYTABKSRVRQ-ASMJPISFSA-N |
Synonym | alpha-maltose, maltose, starch, soluble, glcalpha1-4glca, unii-15sug9ad26, glcalpha1-4glcalpha, amylodextrin, starch solution, alpha-malt sugar, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose |
PubChem CID | 439341 |
ChEBI | CHEBI:18167 |
IUPAC Name | (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol |
SMILES | C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15598194
|
Avantor J.T.Baker
4006-01 |
500 g | Poly Bottle |
216.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15508204
|
Avantor J.T.Baker
4006-05 |
2.5 kg | Poly Bottle |
895.00 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- For Iodometry
Chemical Identifiers
9005-84-9 | |
342.297 | |
alpha-maltose, maltose, starch, soluble, glcalpha1-4glca, unii-15sug9ad26, glcalpha1-4glcalpha, amylodextrin, starch solution, alpha-malt sugar, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose | |
CHEBI:18167 | |
C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O |
C12H22O11 | |
GUBGYTABKSRVRQ-ASMJPISFSA-N | |
439341 | |
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol |
Specifications
9005-84-9 | |
1L = 1.50kg | |
GUBGYTABKSRVRQ-ASMJPISFSA-N | |
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol | |
439341 | |
BAKER ANALYZED™ ACS Reagent | |
Starch, Soluble Potato |
C12H22O11 | |
alpha-maltose, maltose, starch, soluble, glcalpha1-4glca, unii-15sug9ad26, glcalpha1-4glcalpha, amylodextrin, starch solution, alpha-malt sugar, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose | |
C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O | |
342.297 | |
CHEBI:18167 | |
Powder |