missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Starch Solution, Honeywell Fluka™
Indicator, 1% in H2O
350.78 € - 388.26 €
Specifications
Chemical Name or Material | Starch Solution |
---|---|
Density | 1.005g/mL at 25°C |
Synonym | water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Starch Solution | |
1.005g/mL at 25°C | |
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide |