missing translation for 'onlineSavingsMsg'
Learn More

Starch, Thermo Scientific Chemicals

35.07 € - 86.13 €

Specifications

Chemical Name or Material Starch
CAS 9005-84-9, 7732-18-5
Physical Form Solution
Boiling Point 100.0°C
Molecular Formula (C6H10O5)n
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10483543
View Documents
Thermo Scientific Acros
380641000
100 mL Plastic bottle
35.07 €
100mL
In Stock
to see stock.
Edge
Add to basket
 
10639504
View Documents
Thermo Scientific Acros
380645000
500 mL Plastic bottle
86.13 €
500mL
In Stock
to see stock.
Add to basket
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Starch
Solution
(C6H10O5)n
1.0000g/mL
Plastic bottle
1
9005-84-9, 7732-18-5
100.0°C
MFCD00082026
Pure
Passes Test
Videos
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.