Learn More
IBA Lifesciences StarGate pPSG-IBA Acceptor Vectors for E. coli Expression
Achieves high-level expression in E. coli by the T7 promoter by providing specified genetic surroundings for optimal expression and purification of non-toxic proteins.
Brand:  IBA Lifesciences 5-4243-001
 View more versions of this product
Product Code. 15603768
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
pPSG-IGA E. coli expression vectors:
pPSG-IBA vectors use the T7 promoter and T7 RNA polymerase for high-level transcription of the gene of interest.
  • As the T7 promoter is stronger than the tet promoter, pPSG-IBA vectors are recommended in cases where expression with the tet promoter does not lead to significant yields of the recombinant protein.
    • High-level expression in E. coli by bacteriophage T7 promoter
    • High-level transcription by T7 RNA polymerase in BL21 strains
    • High-level expression of non-toxic proteins
    • Induction by IPTG
    • Suitable for in vitro transcription/translation
    StarGate System
    IBA developed StarGate to provide a simple, fast and highly efficient cloning process for genes of interest (GOI) for use in functional expression, purification, detection, immobilization or separation of recombinant proteins:
    • Minimal extra modification of the gene due to short combinatorial sites
    • Inherent high-level cloning efficiency due to a directed reaction (no equilibrium)
    • GOIs are cloned to obtain a Donor Vector; 2 or more genes can be combined using Fusion Cloning
    • Genes are then transferred into Acceptor Vectors that provide optimal genetic surroundings
    • The final expression vector, the Destination Vector, is then placed into the respective host
  • Specifications
    Specifications
    Liquid
    pPSG-IBA 43
    8
    Acceptor Vector for E. coli expression
    2°C to 8°C for frequent usage, -20°C for long-term storage
    250 ng/μL
    Total length: 3469
    Cloning genes of interest for use in functional expression, purification, detection, immobilization or separation of recombinant proteins
    StarGate™ System, E. Coli BL21(DE3)
    E. coli host with T7 RNA polymerase gene (e.g. BL21(DE3))
    T7
    20 μL TE buffer: 10 mm Tris-HCl, 1 mm EDTA / 5 μg
    Strep-tag for C-terminal and His Tag (6x) for N-terminal
    5 μg
    Clear