missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBA Solutions For Life Science StarGate™ Upstream Fusion Vector pNFUSE-IBA
Shop All IBA Lifesciences Products
Click to view available options
Vector:
pNFUSE-IBA w/ IRES11
pNFUSE-IBA w/ LINK11
pNFUSE-IBA w/ LINK12
pNFUSE-IBA w/ SD11
Unit Size:
5µg
Description
StarGate™ offers a simple cloning procedure for rapid and highly efficient subcloning of an arbitrary gene of interest (GOI) for use in functional expression, purification, detection, and immobilization or separation of recombinant proteins.
- Minimal extra modification of the gene due to short combinatorial sites
- Inherent high-level cloning efficiency due to a directed reaction (no equilibrium)
- Genes are cloned in a Donor Vector and then parallel-transfered into Acceptor Vectors that provide different genetic surroundings
- Easily connect two or more genes that are cloned in separate Donor Vectors using Fusion Vectors that are assembled by use of a one-tube reaction
- The GOI to be positioned upstream is transferred into a pNFUSE-IBA derivative, specifying the intergenic region (IR):
- IRES11: Provides internal ribosomal entry site (IRES) from encephalomyocarditis virus for separate cap-independent translation of downstream cistron in mammalian cells
- SD11: IR includes a Shine Dalgarno (SD) sequence for translation of downstream cistron at comparable level to the upstream cistron in bacterial cells
- LINK11: Genes of interest are fused by a sequence encoding the 10 amino acid linker GlySerGlyGlyGlySerGlyGlyGlySer
- LINK12: Genes of interest are fused by a sequence encoding the 15 amino acid linker GlySerGlyGlySerGlyGlyGlyGlySerGlyGlyGlyGlySer
- The GOIs to be positioned downstream are transferred into a pCFUSE-IBA11 vector (identical, irrespective of IR)
Specifications
Specifications
| Format | Solution, liquid |
| For Use With (Equipment) | Stargate™ system derivatives |
| Description | Resistance: ampicillin |
| Includes | IR: A sequence encoding the 15 amino acid linker GlySerGlyGlySerGlyGlyGlyGlySerGlyGlyGlyGlySer |
| pH | 8 |
| Product Type | Fusion Vector |
| Storage Buffer | 20μL TE buffer: 10mM Tris-HCl, 1mM EDTA per 5μL pCFUSE-IBA11 |
| Content And Storage | 2°C to 8°C for frequent usage, -20°C for long-term storage |
| Concentration | 250 ng/μL |
| Quantity | 5 μg |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction