missing translation for 'onlineSavingsMsg'
Learn More

R&L Enterprises™ Stand Rings with Bosshead

Provides for ease of connection on ring stands.

19.95 € - 27.95 €

Specifications

Material Steel
Includes With Bosshead
For Use With (Application) For Rods
View More Specs

Products 3
Product Code Brand Diameter (Metric) Inner Price Quantity & Availability  
Product Code Brand Diameter (Metric) Inner Price Quantity & Availability  
11729037
View Documents
R&L Enterprises™
2013/6
75 mm
19.95 €
Each
In Stock
to see stock.
Edge
Add to basket
 
10796945
View Documents
R&L Enterprises™
2013/11
100 mm
27.95 €
Each
In Stock
to see stock.
Add to basket
 
12996836
View Documents
R&L Enterprises™
2013/14
130 mm
20.55 €
Each
Estimated Shipment: 02-05-2025
to see stock.
Add to basket
 
Description

Description

  • Bosshead with plastic-headed clamping screw
  • Accepts rods from 8mm to 12.5mm in diameter
  • Dimensions given are internal diameter of ring x center to center distance from bosshead to ring
Specifications

Specifications

Steel
With Bosshead
For Rods
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
R&L Enterprises™ Stand Rings with Bosshead

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.