missing translation for 'onlineSavingsMsg'
Learn More

Staurosporine, Fisher BioReagents

Product Code. 10042804
Click to view available options
Quantity:
1 mg
100 μg
500 μg
Packaging:
Poly Micro Tube
This item is not returnable. View return policy
This item is not returnable. View return policy
Staurosporine is a potent protein kinase inhibitor useful for studies on protein phosphorylation in the regulation of cellular functions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62996-74-1
Molecular Formula C28H26N4O3
Molecular Weight (g/mol) 466.541
InChI Key HKSZLNNOFSGOKW-ZYSRIHRCSA-N
Synonym staurosporine, kinome_3629
PubChem CID 49831000
SMILES CC12C(C(CC(O1)N3C4=CC=CC=C4C5=C6C(=C7C8=CC=CC=C8N2C7=C53)CNC6=O)NC)OC

Specifications

Melting Point 270°C
Color White
Assay Percent Range > 98.0 %
Packaging Poly Micro Tube
Quantity 100 μg
Merck Index 15, 8928
Residual Solvents Pass Test
Formula Weight 466.5
Percent Purity >98.0%
Physical Form Solid
Chemical Name or Material Staurosporine, From Streptomyces sp.
Show More Show Less
compliance-icons
Identifiant produit
  • Staurosporine
Mention d'avertissement
  • Warning
Catégorie de danger
  • Carcinogenicity Category 2
Mention de danger
  • H351-Suspected of causing cancer.
Conseils de prudence
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'