Learn More
Invitrogen™ Stealth RNAi™ siRNA Negative Control Med GC Duplex #2
Stealth RNAi™ siRNA Negative Control Med GC Duplex #2
Brand: Invitrogen™ 12935112
251.34 EUR valid until 2025-03-21
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
Stealth RNAi™ siRNA Negative Controls are a great way to measure the effect of your experimental Stealth RNAi™ siRNA duplex versus background. These controls are:Negative Control Kit contains all three controls (Lo, Med, Hi GC content-excluding sequences #2 and #3) and each is also available separately. Get the ideal negative controls for your RNAi experiment so you can assess your results accurately.

Specifications
This individual control is supplied as 250 μl at 20 μM. Store at -20°C. Products are guaranteed stable for 6 months when properly stored. | |
siRNA | |
Unconjugated | |
siRNA | |
Approved for shipment on Wet or Dry Ice |
Negative Control | |
RNAi, Transfection | |
Stealth RNAi™ siRNA | |
250 μL |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2478523 | Certificate of Analysis | 29/01/2025 | 12935112 |
3022531 | Certificate of Analysis | 11/11/2024 | 12935112 |
2263019 | Certificate of Analysis | 07/04/2021 | 12935112 |
2133439 | Certificate of Analysis | 29/09/2019 | 12935112 |
2021526 | Certificate of Analysis | 26/12/2018 | 12935112 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.