missing translation for 'onlineSavingsMsg'
Learn More

Stearic acid, 97%

Product Code. 10722221 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
Fibre drum
Plastic Bottle
Unit Size:
10kg
1kg
2.5kg
This item is not returnable. View return policy

Product Code. 10722221

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 57-11-4 | C18H36O2 | 284.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-11-4
Molecular Formula C18H36O2
Molecular Weight (g/mol) 284.48
MDL Number MFCD00002752
InChI Key QIQXTHQIDYTFRH-UHFFFAOYSA-N
Synonym stearic acid, stearophanic acid, n-octadecanoic acid, cetylacetic acid, pearl stearic, stearex beads, octadecansaeure, stearinsaeure, vanicol, 1-heptadecanecarboxylic acid
PubChem CID 5281
ChEBI CHEBI:28842
IUPAC Name octadecanoic acid
SMILES CCCCCCCCCCCCCCCCCC(O)=O

Specifications

Melting Point 67.0°C to 69.0°C
Color White
Density 0.8400g/mL
Boiling Point 361.0°C
Flash Point 196°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic Bottle
Linear Formula CH3(CH2)16CO2H
Quantity 2.5 kg
Beilstein 02,IV,1206
Merck Index 15,893
Specific Gravity 0.84
Solubility Information Solubility in water: slightly soluble. Other solubilities: 1g/21mL alcohol- 1g/6mL ccl4,1g/5mL benzene- 1g/3.4mL cs2,1g/2mL chloroform - 1g/26mL acetone,soluble in amyl acetate and toluene
Sublimation Point >90 °C
Formula Weight 284.47
Percent Purity 97%
Physical Form Granular Powder, Granules, or Flakes
Chemical Name or Material Stearic acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.