missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kimberly-Clark™ Steel Blue Wypall X80 Cloth
Specifications
Product Type | Wypall X80 Cloth |
---|---|
Material | Non-woven Polyproylene Cellulose Fabric |
Color | Steel Blue |
For Use With (Application) | For heavy-duty tasks in industrial areas, wiping machinery and parts |
Packaging Quantity | 80 Pk. |
Specifications
Wypall X80 Cloth | |
Steel Blue | |
80 Pk. |
Non-woven Polyproylene Cellulose Fabric | |
For heavy-duty tasks in industrial areas, wiping machinery and parts | |
1-ply |