Learn More
Bochem™ Steel Opening Tong
Made from 18/10 stainless steel. The Bochem Steel Opening Tong can open different sized caps.
Brand:  Bochem™ 12965
 View more versions of this product
Product Code. 10729451
  • 350.00 € / Each
Estimated Shipment: 10-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Opening tong
  • For aluminum caps
  • Made from 18/10 stainless steel
Specifications
Specifications
8 mm
Steel