missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Steel Safety Canister

The Steel Safety Canister is ideal for the safe distribution and dosing of flammable liquids.

Brand:  asecos™ 36482

 View more versions of this product

Product Code. 17526646

  • 119.00 € / Each
Estimated Shipment: 09-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Incl. multilingual safety sticker for content labelling, safe distribution and dosing of flammable liquids, with fine dosing tap made of stainless steel (1.4301)
  • Incl. ventilation, NBR seals, integrated flame arrester and plastic protective ring
Specifications

Specifications

2.6 gal.
Yellow
6.69 in.
11.02 in.
6.61 lb.
10 L
Steel
170.00 mm
280.00 mm
3 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers