Learn More
Justrite™ Steel Safety Can for Flammable Liquids
Brand:  Justrite™ 7110100Z
Product Code. 12663135
  • 75.00 € / Each
Estimated Shipment: 13-12-2024
to see stock.
Explore more special offers
Add to basket
Specifications
FM, TUV, UL, ULC, CARB, NFPA, OSHA
4 L
carrying and pouring small quantities of flammable liquids
Powder Coat
241 mm
Safety Can
Red
Steel
sealed lid with automatic positive-pressure relief that vents 0.2 to .35 bar pressure and 40mm fill/pour hole, stainless steel fittings and flame arrester, swinging handle