Learn More
Bochem™ Steel Stand Base
Manufactured according to DIN 12892. Bochem™ Steel Stand Base features M10 thread.
Brand:  Bochem™ 5034
 View more versions of this product
Product Code. 10411203
  • 48.20 € / Each
Estimated Shipment: 31-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Steel
  • Manufactured according to DIN 12892
  • M10 thread
  • One adjustable foot
  • Varnished
Specifications
Specifications
Steel
315 mm
M10 thread
M10
1 Ea.
6 mm
200 mm
Stand Base
Green
3 kg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products