missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Steel Transport Canister

The Steel Transport Canister is ideal to safely transport of liquid substances of packing groups I, II and III.

53.00 € - 80.00 €

Specifications

Color Yellow
Material Steel
View More Specs

Products 5
Product Code Brand Weight (Metric) Price Quantity & Availability  
Product Code Brand Weight (Metric) Price Quantity & Availability  
17886326
View Documents
asecos™
36472
2 kg
53.00 €
Each
Estimated Shipment: 09-01-2025
to see stock.
Add to basket
 
17846386
View Documents
asecos™
36473
2.2 kg
53.00 €
Each
Estimated Shipment: 09-01-2025
to see stock.
Add to basket
 
17806406
View Documents
asecos™
36478
2.75 kg
62.00 €
Each
Estimated Shipment: 09-01-2025
to see stock.
Add to basket
 
17886386
View Documents
asecos™
36474
3.3 kg
73.00 €
Each
Estimated Shipment: 09-01-2025
to see stock.
Add to basket
 
17876326
View Documents
asecos™
36479
3.8 kg
80.00 €
Each
Estimated Shipment: 09-01-2025
to see stock.
Add to basket
 
Description

Description

  • Incl. multilingual safety sticker for content labelling, with aluminium screw cap, ventilation valve, NBR seals and plastic protective ring
  • UN-approval for packaging groups I, II and III
  • Steel
Specifications

Specifications

Yellow
Steel
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.