missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bochem™ Steel Wire Mesh
3.10 € - 5.65 €
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | ||||||
10410353
|
Bochem™
12751 |
120 x 120 mm |
3.10 €
Each |
Estimated Shipment: 20-06-2025
Log in to see stock. |
||||||
10172361
|
Bochem™
12753 |
200 x 200 mm |
5.65 €
Each |
Estimated Shipment: 20-06-2025
Log in to see stock. |
||||||
Specifications
Wire Gauze with Ceramic Center | |
Square |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Bochem™ Steel Wire Mesh