missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Biosciences™ Stem Cell Control CD34+ Whole Blood Process Control (IVD)
missing translation for 'orderingAttributeHoverText'
Quantity:
1 x 2.0 mL CD34+ Low, 1 x 2.0 mL CD34+ High
missing translation for 'unitSize'
Each
Descripción
- Complete, two-level, process control for immunophenotyping and enumeration of leukocytes by flow cytometry.
- Control for antibody staining, red blood cell (RBC) lysis, instrument setup and performance, and data analysis.
Especificaciones
Especificaciones
| Content And Storage | Store vials upright, tightly capped, at 2°C to 8°C when not in use. |
| Concentration | CD34+ Low approx. 10 cells/μL, CD34+ High approx. 35 cells/μL |
| For Use With (Equipment) | Flow cytometer |
| Intended Use | Complete, two-level, process control for immunophenotyping and enumeration of leucocytes by flow cytometry. |
| Quantity | 1 x 2.0 mL CD34+ Low, 1 x 2.0 mL CD34+ High |
| Regulatory Write-In | IVD |
| Stability | Unopened vials are stable until the expiration date indicated on each vial and assay sheet. Opened vials are stable for 12 thermal cycles (uses) when handled properly. Avoid unnecessary cycles of warming and cooling. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido