Learn More
Thermo Scientific Chemicals Stem-Cell Factor/c-Kit Inhibitor
Brand:  Thermo Scientific Chemicals J65114.MF
 View more versions of this product
Product Code. 15486729
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Inhibits in vitro phosphorylation of c-Kit. Also inhibits SCF (stem cell factor)-induced c-kit phosphorylation in 501mel human melanoma cells
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
355.50
Plastic bottle
Solid
50mg
Light sensitive