Learn More
STEMCELL Technologies STEMdiff™ NK Cell Kit
For expansion and differentiation of hPSCs to NK cells
Brand:  STEMCELL Technologies 100-0170
Encompass
Product Code. 17177491
  • 2202.00 € / 1 set
Explore more special offers
Description
Description
  • STEMdiff™ NK Cell Kit includes serum-free media and supplements for the feeder-free differentiation of human pluripotent stem cells (PSCs) to natural killer (NK) cells expressing CD56
  • STEMdiff™ Hematopoietic - EB reagents are animal-component free and optimized for lymphoid differentiation potential
  • Using these reagents, CD34+ hematopoietic progenitor cells are first generated from PSCs; these can then be differentiated to NK cells using the StemSpan™ components
Specifications
Specifications
Hematopoietic Cells, PSC-Derived, NK Cells, Pluripotent Stem Cells
Cell Culture, Differentiation, Expansion
1 Kit