Learn More
Zeiss™ Stemi 305 Stereo Microscope
Compact Greenough stereo microscope with 5:1 zoom for biological education, labs and production environments.
1960.00 € - 2205.00 €
Specifications
Product Type Stereo Microscope
View More Specs
Products 2
Product Code Brand Includes Price Quantity & Availability  
Product Code Brand Includes Price Quantity & Availability  
15986182
View Documents
Zeiss™
435063-9020-100
Double Spot Illuminator K LED
2205.00 €
Each
Estimated Shipment: 18-02-2025
to see stock.
Add to basket
17332810
View Documents
Zeiss™
435063-9030-100
Ring Illuminator K LED
1960.00 €
1 set
Estimated Shipment: 18-02-2025
to see stock.
Add to basket
Description
Description
  • This compact Greenough stereo microscope allows you to observe your samples as they are: three-dimensional and crisp in contrast and no preparation required.
  • The Stemi 305 is an easy-to-use microscope, requires minimal setup, everything is integrated including long-living LED illumination for both reflected and transmitted light.
  • Spot Illuminator K LED - Single spot for brightfield and darkfield epi-illumination, Thread M24x0.5 for polarizer, Height adjustable, inclinable, zoomable.
Specifications
Specifications
Stereo Microscope