missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ StemPro™ Chondrogenesis Differentiation Kit

StemPro™ Chondrogenesis Differentiation Kit

Brand:  Gibco™ A1007101

182.40 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.


Product Code. 10771764

  • 205.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The StemPro™ Chondrogenesis Differentiation Kit has been developed for the chondrogenic differentiation of mesenchymal stem cells (MSCs) in tissue-culture vessels. The kit contains all reagents required for inducing MSCs to be committed to the chondrogenesis pathway and generate chondrocytes. Using StemPro™ Chondrogenesis Differentiation Kit in combination with StemPro™ MSC SFM or MesenPRO RS™ Medium provides a standardized culture workflow solution for MSC isolation, expansion, and differentiation into collagen matrix-producing chondrocytes.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Stem Cells (Mesenchymal)
CO2
Chondrogenesis Differentiation Kit
Liquid
GlutaMAX™-I
No Phenol Red
Research Grade
Differentiation
Sodium Bicarbonate
Human
This kit is shipped with a 100 ml basal medium and 10 ml growth supplement adequate to formulate 100 ml of complete medium. Components require separate shipping and storage requirements. The growth supplement is not sold individually.
StemPro™ Osteocyte⁄Chondrocyte Differentiation Basal Medium 2 to 8°C (protect from light)
StemPro™ Chondrogenesis Supplement -5 to -20°C (in the dark)
Stem Cell Differentiation
≤50 EU/mL, ≤ 50 EU/ml
Low Glucose
No HEPES
Gibco™, StemPro™
1 Kit
Serum-Free
Sodium Pyruvate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.