missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ StemPro™ MSC SFM XenoFree
StemPro™ MSC SFM XenoFree
Brand: Gibco™ A1067501
440.97 EUR valid until 2024-12-31
Use promo code "21952" to get your promotional price.
This item is not returnable.
View return policy
Description
GIBCO™ StemPro™ MSC SFM XenoFree has been developed for the growth and expansion of human mesenchymal stem cells (MSCs) and Adipose-derived Stem Cells (ADSCs) under completely serum-free and xenogeneic (xeno)-free conditions. Using StemPro™ MSC SFM XenoFree, human MSCs or ADSCs can be expanded for multiple passages while maintaining their multipotent phenotype (i.e. ability to differentiate into osteogenic, chondrogenic and adipogenic lineages). StemPro™ MSC SFM XenoFree is cGMP compliant which allows for traceability and manufacturing reliability.Specifications
Each kit contains 1 x 500 ml StemPro™ MSC SFM Basal Medium and 1 X 5 ml StemPro™ MSC SFM XenoFree Supplement. StemPro™ MSC SFM Basal Medium is stored at 2 to 8°C and stable for 12 months. The StemPro™ MSC SFM XenoFree Supplement is stored frozen at -5 to -20°C and stable for 12 months. | |
Mesenchymal Stem Cells | |
Adherent Cell Culture | |
Stem Cell Serum Free Media (SFM) | |
No Glutamine, No Phenol Red | |
StemPro™ | |
1 kit |
CO2 | |
Xeno-free | |
Liquid | |
Serum-free | |
No antibiotics | |
MS Cell-Qualified | |
Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction