missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ StemPro™ NSC SFM

StemPro™ NSC SFM

Brand:  Gibco™ A1050901

403.75 EUR valid until 2025-12-31
Use promo code "24342" to get your promotional price.


Product Code. 10377893

  • 457.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Neural Stem Cell Serum Free Medium that provides superior expansion of human NSC's compared to competitive products.

  • Supports differentiation capability into astrocytes, oligodendrocytes and neurons
  • Supports adherent or spheroid cultures
  • Developed for Human Neural Stem Cell Culture

    StemPro NSC SFM is a serum-free medium (SFM) specially formulated for the growth and expansion of human neural stem cells (NSCs). StemPro™ NSC SFM enables superior human NSC expansion of both adherent and spheroid cultures derived from human embryonic stem cells or fetal tissue compared to existing commercial products. StemPro NSC SFM also maintains the multipotential differentiation capability into astrocytes, oligodendrocytes and neurons. Each lot of StemPro™ NSC SFM is cGMP manufactured and qualified using a human NSC proliferation assay.
    TRUSTED_SUSTAINABILITY
  • Specifications

    Specifications

    Neural Stem Cells
    Liquid
    Serum-free
    No HEPES
    1 Kit
    ≤50 EU/mL
    Research Grade
    This kit is shipped with a basal medium, growth supplement, and growth factors adequate to formulate 500 ml of complete medium. The components require separate shipping and storage requirements:
    KnockOut™ DMEM⁄F-12 Basal Medium (500 mL; store in the dark 2 to 8°C)
    StemPro™ NSC SFM Supplement (10 mL; store frozen at -5 to -20°C)
    FGF Basic Recombinant Human and EGF Recombinant Human (10 ug; store at 2 to 8°C desiccated).
    Mammalian Cell Culture
    Serum-free
    Stem Cell Serum Free Media (SFM)
    Low Glucose, Glutamine, Phenol Red, Sodium Pyruvate, Sodium Bicarbonate
    Gibco™, StemPro™
    Non-Essential Amino Acids
    Sodium Phosphate
    Human
    CO2
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title
    Gibco™ StemPro™ NSC SFM > 1 kit

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

    For Research Use Only. Not for use in diagnostic procedures.