missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ StemPro™ Osteogenesis Differentiation Kit
StemPro™ Osteogenesis Differentiation Kit
Brand: Gibco™ A1007201
186.20 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.
This item is not returnable.
View return policy
Description
For Complete Differentiation of Human Mesenchymal Stem Cells into Bone CellsStemPro™ Osteogenesis Differentiation Kit has been developed for the osteogenic differentiation of mesenchymal stem cells (MSCs) in tissue culture vessels. The kit contains all reagents required for inducing MSCs to be committed to the osteogenesis pathway and generate
immature and mature osteocytes. Using StemPro™ Osteogenesis Differentiation Kit in combination with MesenPRO RS™ Medium or StemPro™ MSC SFM provides a standardized culture workflow solution for MSC isolation, expansion and differentiation into mineralized matrix-producing osteocytes.
Specifications
Stem Cells (Mesenchymal) | |
CO2 | |
Osteogenesis Differentiation Kit | |
Liquid | |
1000 mg/L | |
No HEPES | |
No Phenol Red | |
Research Grade | |
Differentiation | |
Sodium Bicarbonate | |
Human |
This kit is shipped in two parts with separate storage requirements. These components are not sold individually: • StemPro™ Osteocyte/Chondrocyte Differentiation Basal Medium (100 ml; store in the dark at 2°C to 8°C) • StemPro™ Osteogenesis Supplement (10 ml; store in the dark at -5°C to -20°C) The Basal Medium ships at room temperature, while the Supplement ships on dry ice. |
|
Stem Cell Differentiation | |
≤50 EU/mL, ≤ 50 EU/ml | |
Low Glucose | |
GlutaMAX™-I | |
Sodium Phosphate | |
Gibco™, StemPro™ | |
1 Kit | |
Standard Serum | |
Sodium Pyruvate | |
Osteogenesis Supplements |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction