Learn More
R&D Systems™ StemXVivo Adipogenic Supplement (100X)
Media supplement to induce the differentiation of human, mouse or rat MSCs into adipocytes
Brand:  R&D Systems™ CCM011
Product Code. 18772464
  • 233.00 € / 3.50mL
Explore more special offers
Description
Description
Despite the well-characterized factors and protocols used to differentiate mesenchymal stem/stromal cells (MSCs) into adipocytes, differentiation efficiencies can vary depending on the quality of the MSC starting population and the reagents used to expand and differentiate MSCs.
  • Contains high quality differentiation factors to drive reproducible and efficient MSC adipogenesis
  • Is defined to reduce unwanted experimental variability
  • Has been developed and optimized using MSCs
Specifications
Specifications
Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
3.5 mL