Learn More
R&D Systems™ StemXVivo Hepatocyte Differentiation Kit
Uses high-quality specialized media and pre-mixed differentiation cocktails to maximize differentiation efficiency and ensure the consistent and reliable generation of scalable amounts of hepatocyte-like cells
Brand:  R&D Systems™ SC033
562.41 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 30001962
  • 714.00 € / Each
Explore more special offers
Description
Description
  • Optimized components ensure highly enriched populations of hepatocyte-like cells
  • Reproducible differentiation protocols translate into cost and time savings
  • Maximizes workflow efficiency by standardizing hepatocyte differentiation
  • Qualified for hepatotoxic drug and small molecule screening
Specifications
Specifications
  • Hepatocyte Differentiation Cocktail I
  • Hepatocyte Differentiation Cocktail II
  • Hepatocyte Differentiation Cocktail III
  • Hepatocyte Differentiation Cocktail IV
  • Hepatocyte Differentiation Base Media I
  • Hepatocyte Differentiation Base Media II
  • Anti-Human Serum Albumin
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
StemXVivo Hepatocyte Differentiation Kit
1 kit
For differentiation of pluripotent stem cell into hepatocytes