missing translation for 'onlineSavingsMsg'
Learn More

Merck Stericup™ Quick Release-GP Sterile Vacuum Bottle Top Filtration Systems

The Stericup™ vacuum filtration system is the most efficient single-use, disposable Bottle Top Filter process for sterile media preparation. It combines a Steritop™ filter unit with a receiver flask for processing and storing volumes from 150 mL to 1000 mL in a single, ready-to-use format.

193.00 € - 485.00 €

Specifications

Color Clear with blue adapter
Disposable Single-use
Pore Size 0.22 μm
Sterility Sterile
For Use With (Application) Sterile filtration
View More Specs

Products 4
Product Code Brand Volume (Metric) Volume (Metric) Receiver Price Quantity & Availability  
Product Code Brand Volume (Metric) Volume (Metric) Receiver Price Quantity & Availability  
15949180
View Documents
Merck
S2GPU01RE
150 mL 150 mL
193.00 €
Pack of 12
In Stock
to see stock.
Add to basket
 
15959180
View Documents
Merck
S2GPU02RE
250 mL 250 mL
219.00 €
Pack of 12
In Stock
to see stock.
Add to basket
 
15969180
View Documents
Merck
S2GPU10RE
1000 mL 500 mL
462.00 €
Pack of 12
Estimated Shipment: 10-12-2024
to see stock.
Add to basket
 
15979180
View Documents
Merck
S2GPU11RE
1000 mL 1000 mL
485.00 €
Pack of 12
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

The Stericup™ vacuum filtration system is the most efficient single-use, disposable Bottle Top Filter process for sterile media preparation. It combines a Steritop™ filter unit with a receiver flask for processing and storing volumes from 150 mL to 1000 mL in a single, ready-to-use format.

The Stericup™ vacuum filtration system is the most efficient single-use, disposable process for sterile media preparation. It combines a Steritop™ filter unit with a receiver flask for processing and storing volumes from 150 mL to 1000 mL in a single, ready-to-use format.

Its no-tip, easy-grip flask design and balanced profile improve stability during filtration in tight laminar flow hoods, and the convenient labeling and packaging make it easy to open and track each device.

The bottom of the receiver flask is slightly recessed, so you can conveniently stack capped flasks after filtration, maximizing precious refrigeration space. You can also use Steritop™ bottle-top devices with your own sterile capture flask or bottle.

Features & Benefits

  • Millipore Express™ PLUS membrane for fastest flow rates and low protein binding to minimize absorption and protein loss, which affect downstream performance and costs.
  • Durapore™ membrane for ultra-low protein binding for minimum absorption and protein loss.
  • Snug tight packaging, easy tear-open pouch, and convenient label designs make storing, opening and tracking your devices easy and efficient.
  • Stable base and easy grip design of the Stericup™ device enable any user to comfortably perform all stages of the workflow in a laminar flow hood.
  • Recessed bottom of Stericup™ receiver flasks allows flasks to be stacked easily in refrigerated spaces.

Applications

  • Sterilization of Aqueous Solutions (150 mL to 1 L)
    • Sterile Filtration of Culture Media and Buffers with High-Value Protein or Small Molecule Component.
Specifications

Specifications

Clear with blue adapter
0.22 μm
Sterile filtration
73 mm
Single-use
Sterile
Bottle Top Filter
Videos
SDS
Documents

Documents

Product Certifications