Learn More
Merck Stericup™ Quick Release-HV Sterile Vacuum Bottle Top Filtration Systems
Stericup™ Quick Release-HV Sterile Vacuum Filtration System has a design and balanced profile that improves stability during filtration in tight laminar flow hoods.
182.00 € - 490.00 €
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
15997996
|
Merck
S2HVU01RE |
150 mL |
182.00 €
Pack of 12 |
Estimated Shipment: 11-03-2025
Log in to see stock. |
|||||
15908006
|
Merck
S2HVU02RE |
250 mL |
223.00 €
Pack of 12 |
Estimated Shipment: 30-01-2025
Log in to see stock. |
|||||
15918006
|
Merck
S2HVU05RE |
500 mL |
329.00 €
Pack of 12 |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||||
15928006
|
Merck
S2HVU11RE |
1000 mL |
490.00 €
Pack of 12 |
Estimated Shipment: 11-03-2025
Log in to see stock. |
|||||
Description
Stericup™ Quick Release-HV Sterile Vacuum Filtration System has a design and balanced profile that improves stability during filtration in tight laminar flow hoods.
The Stericup™ vacuum filtration system is the most efficient single-use, disposable process for sterile media preparation. It combines a Steritop™ filter unit with a receiver flask for processing and storing volumes from 150 mL to 1000 mL in a single, ready-to-use format.
Its no-tip, easy-grip flask design and balanced profile improve stability during filtration in tight laminar flow hoods, and the convenient labeling and packaging make it easy to open and track each device.
The bottom of the receiver flask is slightly recessed, so you can conveniently stack capped flasks after filtration, maximizing precious refrigeration space. You can also use Steritop™ bottle-top devices with your own sterile capture flask or bottle.
Features & Benefits
- Millipore Express™ PLUS membrane for fastest flow rates and low protein binding to minimize absorption and protein loss, which affect downstream performance and costs.
- Durapore™ membrane for ultra-low protein binding for minimum absorption and protein loss.
- Snug tight packaging, easy tear-open pouch, and convenient label designs make storing, opening and tracking your devices easy and efficient.
- Stable base and easy grip design of the Stericup™ device enable any user to comfortably perform all stages of the workflow in a laminar flow hood.
- Recessed bottom of Stericup receiver flasks allows flasks to be stacked easily in refrigerated spaces.
Applications
- Sterilization of Aqueous Solutions (150 mL to 1 L)
- Sterile Filtration of Culture Media and Buffers with High-Value Protein or Small Molecule Component.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.