Learn More
Merck Stericup™ Quick Release Bottle Top Receiver Flasks
Stericup™ Quick Release Receiver Flask for Bottle Top Filtration includes a click-seal confidence cap that enables a secure grip in wet and dry conditions.
87.00 € - 176.00 €
Specifications
Outlet Fitting 45 mm
View More Specs
Products 3
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
15967996
View Documents
Merck
S200B02RE
250 mL
87.00 €
Pack of 12
Estimated Shipment: 07-03-2025
to see stock.
Add to basket
15977996
View Documents
Merck
S200B05RE
500 mL
112.00 €
Pack of 12
Estimated Shipment: 18-03-2025
to see stock.
Add to basket
15987996
View Documents
Merck
S200B10RE
1000 mL
176.00 €
Pack of 12
Estimated Shipment: 05-03-2025
to see stock.
Add to basket
Description
Description
Stericup™ Quick Release Receiver Flask for Bottle Top Filtration includes a click-seal confidence cap that enables a secure grip in wet and dry conditions.
Steritop™ bottle top filter units provide a large filter area for rapid sterile filtration of aqueous fluids into narrow-mouth (33 mm) or wide-mouth (45 mm) bottles.
A small tab inside the funnel secures an optional prefilter in-place during filtration.
For added protection against contamination, the threads for bottle attachment have been slightly recessed in case the funnel accidentally touches a work surface.
Specifications
Specifications
45 mm
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Merck Stericup™ Quick Release Bottle Top Receiver Flasks