Learn More
Merck Millipore Sterikon™ plus Bioindicator
For monitoring the effectiveness of steam sterilization. Sterikon™ plus Bioindicator ampules contain spores of an apathogenic organism that is destroyed with sufficient autoclaving. When the ampoules are incubated, incomplete sterilization is indicated by growth of the organism.
Brand:  Merck Millipore 1102740002
Product Code. 10036120
  • 700.00 € / 100 ampoules
Explore more special offers
Description
Description
The Sterikon™ plus Bioindicator System is used to monitor the effectiveness of steam sterilization at 121°C for 15 minutes. It consists of an ampule that contains nutrient broth, sugar, a pH indicator and spores of an apathogenic organism, Geobacillus stearothermophilus ATCC 7953 (sporulation optimized).
  • Spores are totally killed after 15 min. when heated at 121°C ± 0.5°C (1 bar)
  • At lower temperatures or lower exposure times, the spores can survive, at least partly
  • An appropriate number of ampules are placed into the autoclave along with the batch to be sterilized
  • The ampules are incubated up to 48 hours at 60 ± 2°C
  • No growth of the organism indicates adequate sterilization; growth shows inadequate sterilization