missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Contec™ Sterile Denatured Ethanol, 70%
Blend of 70% v/v denatured ethanol with 30% purified water. Contec™ Sterile Low Endotoxin Alcohol, 70% is 0.2 micron filtered.
Brand: Contec™ SBC570DE
160.07 EUR valid until 2024-09-30
Use promo code "21675" to get your promotional price.
This item is not returnable.
View return policy
Description
- Guaranteed endotoxin levels of <0.25EU/mL make it ideal for use in product contact areas
- GMP manufactured under Grade A laminar flow air flow in a Grade C cleanroom ensures the alcohol, container and packaging are free from contamination and particulates
- Trigger spray and bag in bottle protected system prevents bottle contents from becoming contaminated during use
- Bottles can be completely emptied eliminating wastage so no need to discard unused product during shelf life
- Trigger spray can be set to jet or spray - large droplet size reduces the risk of inhalation and provides good surface coverage
- Double/triple bagged packed in linear tear packaging so each bag is easy to open even when wearing gloves
- Sterile via gamma irradiation so suitable for use in Grade A and B cleanrooms
Specifications
Denatured Ethanol | |
Not Applicable | |
Colorless | |
70% denatured ethanol with purified water | |
Capped Bottle | |
Limited-use | |
Yes | |
5 L |
Liquid | |
Sterile | |
Clear | |
70% (v/v) Ethanol in 30% Purified Water | |
Sterilised by gamma irradiation at no less than 25 kGy | |
Filtered | |
2 x 5 L |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- contec sterile denatured ethanol
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P370+P378-In case of fire: Use dry chemical/carbondioxide to extinguish.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P411-Store at temperatures not exceeding 40°C/°F.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol, propan-2-ol